Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Antibody (7H3D7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP252500 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP252500 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP252500 Supplier Novus Biologicals Supplier No. NBP252500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Monoclonal specifically detects Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.

Specifications

Antigen Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B
Applications Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry
Classification Monoclonal
Clone 7H3D7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready
Formulation PBS with 0.05% Sodium Azide
Gene Alias CT31FLJ10538, EC 1.14.11, EC 1.14.11.-, homolog 1A, JARID1BFLJ16281, jumonji, AT rich interactive domain 1B, Jumonji, AT rich interactive domain 1B (RBP2-like), lysine (K)-specific demethylase 5B, PLU-1FLJ12459, PLU1FLJ23670, PUT1, RBBP2H1, RBBP2H1AFLJ12491, RBP2-H1, Retinoblastoma-binding protein 2 homolog 1
Gene Symbols KDM5B
Host Species Mouse
Immunogen Purified recombinant fragment of human KDM5B (AA: 231-319) expressed in E. Coli.
Molecular Weight of Antigen 175.6 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Chromatin Modifiers, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 10765
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.