Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB185382 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB185382 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB185382 Supplier Novus Biologicals Supplier No. NBP282107AF405
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Polyclonal antibody specifically detects Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 1.14.11, EC 1.14.11.-, JmjC domain-containing protein 3, JMJD3lysine-specific demethylase 6B, jumonji domain containing 3, histone lysine demethylase, Jumonji domain-containing protein 3, KIAA0346jumonji domain containing 3, lysine (K)-specific demethylase 6B, Lysine demethylase 6B
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 16 amino acids near the carboxy terminus of human Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3. The immunogen is located within amino acids 1370-1420 of Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3. Amino Acid Sequence: KVPGSRTPGHQENNNF
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 23135
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.