Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB183483 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB183483 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB183483 Supplier Novus Biologicals Supplier No. NBP306855AF532
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Polyclonal antibody specifically detects Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias EC 1.14.11, EC 1.14.11.-, JmjC domain-containing protein 3, JMJD3lysine-specific demethylase 6B, jumonji domain containing 3, histone lysine demethylase, Jumonji domain-containing protein 3, KIAA0346jumonji domain containing 3, lysine (K)-specific demethylase 6B, Lysine demethylase 6B
Host Species Rabbit
Immunogen Partial recombinant protein made to an internal portion of the human JMJD3 protein (amino acids 1520-1682) [UniProt O15054]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 23135
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.