Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lysine-Specific Demethylase 4C (JMJD2C) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35
Specifications
Antigen | Lysine-Specific Demethylase 4C (JMJD2C) |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABE1409-25UG
|
MilliporeSigma
ABE140925UG |
25 μg |
Each for $192.35
|
|
|||||
ABE1409MI
|
MilliporeSigma
ABE1409 |
100 μg | N/A | N/A | N/A | ||||
Description
Anti-Lysine-Specific Demethylase 4C (JMJD2C) Antibody is a highly specific rabbit polyclonal antibody that targets Lysine-specific demethylase 4C and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.Specifications
Lysine-Specific Demethylase 4C (JMJD2C) | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse | |
KLH-Conjugated linear peptide corresponding to 18 amino acids from the N-terminal half of human Lysine-specific demethylase 4C. | |
Affinity Purified | |
This polyclonal antibody detects Lysine-specific demethylase 4C in multiple human and mouse cell types. |
Please refer to lot specific datasheet. | |
Polyclonal | |
Rabbit | |
Epigenetics and Nuclear Function | |
KDM4C,GASC1,JHDM3C,JMJD2C,KIAA0780 | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title