Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysozyme Antibody (BGN/0696/5B1), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB10063062X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10063062X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10063062X Supplier Novus Biologicals Supplier No. NB10063062AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lysozyme Monoclonal specifically detects Lysozyme in Human samples. It is validated for CyTOF, ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen Lysozyme
Applications CyTOF, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BGN/0696/5B1
Conjugate Alexa Fluor 488
Dilution Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P61626
Gene Alias EC 3.2.1.17, lysozyme, lysozyme (renal amyloidosis)1,4-beta-N-acetylmuramidase C, lysozyme C, LZM, renal amyloidosis
Gene Symbols LYZ
Host Species Mouse
Immunogen Native, from urine.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4069
Test Specificity NB100-63062 recognizes lysozyme, an enzyme that is present in mucosal secretions such as saliva and tears and is involved with non-specific organism defence. It functions by hydrolyzing 1,2-beta linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues such as those found in the walls of various Gram-positive bacteria.
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.