Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysozyme Antibody (LZ598-10G9), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB002799 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002799 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002799 Supplier Novus Biologicals Supplier No. NBP262223AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lysozyme Monoclonal specifically detects Lysozyme in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen Lysozyme
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone LZ598-10G9
Conjugate Alexa Fluor 700
Dilution Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.2.1.17, lysozyme, lysozyme (renal amyloidosis)1,4-beta-N-acetylmuramidase C, lysozyme C, LZM, renal amyloidosis
Gene Symbols LYZ
Host Species Mouse
Immunogen human lysozyme
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cardiovascular Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4069
Test Specificity The antibody MEM-188 reacts with a 180 kDa isoform of CD56 (NCAM) expressed in leukocytes. It has been suggested that the antibody MEM-188 could react with rhesus monkey lymphocytes. Reactivity with other NCAM isoforms has not been tested.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.