Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysozyme Antibody (OTI1C9), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB288048 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288048 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288048 Supplier Novus Biologicals Supplier No. NBP272557APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lysozyme Monoclonal antibody specifically detects Lysozyme in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Lysozyme
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1C9
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias EC 3.2.1.17, lysozyme, lysozyme (renal amyloidosis)1,4-beta-N-acetylmuramidase C, lysozyme C, LZM, renal amyloidosis
Host Species Mouse
Immunogen Full length human recombinant protein of human LYZ(NP_000230) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4069
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.