Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysozyme, Chicken Egg White PROMO

Catalog No. J6070122F25
Click to view available options
:
100 g

Lysozyme, chicken egg white, CAS # 12650-88-3, is an enzyme that catalyzes the hydrolysis of peptidoglycans found in gram-positive bacterial cell walls, causing bacterial cell lysis.

General Description

  • Lysozyme, chicken egg white is a naturally occurring enzyme that hydrolyzes the peptidoglycans in the bacterial cell wall, resulting in cell wall lysis

Application

  • Lysozyme is utilized for the hydrolysis of peptidoglycans found in bacterial cell walls
  • It is used as a precursor in the production of spheroplasts in laboratory settings
  • It is used for lysing E. coli and Streptomycetes for extraction purposes, such as extracting group-specific antigen
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12650-88-3
Molecular Formula C125H196N40O36S2

Specifications

Color White
Solubility Soluble Aqueous Solution at 10 mg/ml
Content And Storage Store at -20°C
Quantity 100 g
Product Type Lysozyme
Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.