Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lysyl Oxidase Homolog 2/LOXL2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | Lysyl Oxidase Homolog 2/LOXL2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17406520
![]() |
Novus Biologicals
NBP17406520UL |
20 μL |
Each for $206.00
|
|
|||||
NBP174065
![]() |
Novus Biologicals
NBP174065 |
100 μL |
Each for $487.50
|
|
|||||
Description
Lysyl Oxidase Homolog 2/LOXL2 Polyclonal specifically detects Lysyl Oxidase Homolog 2/LOXL2 in Mouse samples. It is validated for Western Blot.Specifications
Lysyl Oxidase Homolog 2/LOXL2 | |
Polyclonal | |
Rabbit | |
P58022 | |
4017 | |
Synthetic peptides corresponding to the C terminal of Loxl2. Immunizing peptide sequence NNGQSDFRPKNGRHAWIWHDCHRHYHSMEVFTYYDLLSLNGTKVAEGHKA. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
EC 1.4.3, EC 1.4.3.-, LOR2, lysyl oxidase homolog 2, lysyl oxidase related 2, lysyl oxidase-like 2, Lysyl oxidase-like protein 2, Lysyl oxidase-related protein 2, Lysyl oxidase-related protein WS9-14, WS9-14 | |
LOXL2 | |
IgG | |
85 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title