Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysyl Oxidase Homolog 2/LOXL2 Rabbit anti-Human, Clone: 9F0W6, Novus Biologicals™
SDP

Catalog No. NB168023 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168023 100 μg
NB168024 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168023 Supplier Novus Biologicals Supplier No. NBP316466100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Lysyl Oxidase Homolog 2/LOXL2 Monoclonal antibody specifically detects Lysyl Oxidase Homolog 2/LOXL2 in Human samples. It is validated for Western Blot

Specifications

Antigen Lysyl Oxidase Homolog 2/LOXL2
Applications Western Blot
Classification Monoclonal
Clone 9F0W6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 1.4.3, EC 1.4.3.-, LOR2, lysyl oxidase homolog 2, lysyl oxidase related 2, lysyl oxidase-like 2, Lysyl oxidase-like protein 2, Lysyl oxidase-related protein 2, Lysyl oxidase-related protein WS9-14, WS9-14
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 675-774 of human Lysyl Oxidase Homolog 2/LOXL2 (Q9Y4K0).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4017
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.