Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysyl Oxidase Homolog 3/LOXL3 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NBP275964F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275964F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275964F Supplier Novus Biologicals Supplier No. NBP275964F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lysyl Oxidase Homolog 3/LOXL3 Polyclonal specifically detects Lysyl Oxidase Homolog 3/LOXL3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Lysyl Oxidase Homolog 3/LOXL3
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 1.4.3, EC 1.4.3.-, EC 1.4.3.13, LOXL, lysyl oxidase homolog 3, lysyl oxidase-like 3, Lysyl oxidase-like protein 3
Gene Symbols LOXL3
Host Species Rabbit
Immunogen Partial recombinant human LOXL3 protein (amino acids 26-753) [UniProt P58215]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 84695.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.