Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysyl tRNA synthetase Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB084981 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB084981 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB084981 Supplier Novus Biologicals Supplier No. NBP299362JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lysyl tRNA synthetase Polyclonal specifically detects Lysyl tRNA synthetase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Lysyl tRNA synthetase
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50 mM Sodium Borate
Gene Alias CMTRIB, EC 6.1.1, EC 6.1.1.6, KARS2, KIAA0070, KRS, lysine tRNA ligase, Lysine--tRNA ligase, LysRS, lysyl-tRNA synthetase
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human Lysyl tRNA synthetase.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 3735
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.