Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysyl tRNA synthetase Antibody, Novus Biologicals™
SDP

Catalog No. NB1001719 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001719 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001719 Supplier Novus Biologicals Supplier No. NB1001719
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lysyl tRNA synthetase Polyclonal specifically detects Lysyl tRNA synthetase in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Lysyl tRNA synthetase
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:250, Immunoprecipitation 1-4 ug/mg of lysate, Immunohistochemistry-Paraffin 1:10-1:500, Immunocytochemistry
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_005539.1
Gene Alias CMTRIB, EC 6.1.1, EC 6.1.1.6, KARS2, KIAA0070, KRS, lysine tRNA ligase, Lysine--tRNA ligase, LysRS, lysyl-tRNA synthetase
Gene Symbols KARS1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 550 and the C-terminus (residue 597) of human Lysyl-tRNA Synthetase using the numbering given in entry NP_005539.1 (GeneID 3735).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 3735
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.