Learn More
Description
Specifications
Specifications
| Antigen | LYVE-1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | ALY7 |
| Conjugate | PE |
| Dilution | Flow Cytometry |
| Gene Alias | cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE-1XLKD1 |
| Gene Symbols | LYVE1 |
| Host Species | Rat |
| Immunogen | This LYVE-1 Antibody (ALY7) was developed against mouse LYVE1. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
