Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYVE-1 Antibody (ALY7), PerCP, Novus Biologicals™
SDP

Catalog No. NBP143411P Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP143411P 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP143411P Supplier Novus Biologicals Supplier No. NBP143411PCP
Add to Cart
Add to Cart

Rat Monoclonal Antibody

LYVE-1 Monoclonal specifically detects LYVE-1 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen LYVE-1
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone ALY7
Conjugate PerCP
Dilution Flow Cytometry
Gene Alias cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE-1XLKD1
Gene Symbols LYVE1
Host Species Rat
Immunogen This LYVE-1 Antibody (ALY7) was developed against mouse LYVE1.
Molecular Weight of Antigen 35 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cardiovascular Biology, Endosome Markers, Endothelial Cell Markers, Immunology, Inflammation, Metastasis
Primary or Secondary Primary
Gene ID (Entrez) 10894
Test Specificity Human reactivity reported in scientific literature (PMID: 23975155).
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.