Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LYVE-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$336.16 - $691.20
Specifications
| Antigen | LYVE-1 |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
LYVE-1 Polyclonal specifically detects LYVE-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| LYVE-1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q9Y5Y7 | |
| 10894 | |
| This LYVE-1 Antibody was developed against a recombinant protein corresponding to amino acids: FETCSYGWVGDGFVVISRISPNPKCGKNGVGVLIWKVPVSRQFAAYCYNSSDTWTNSCIPEIITTKDPIFNTQTATQTTEFIVSDSTYSVASPYSTIPAPTTTPPAPASTSIPRRKKLICVTEVFMETSTMSTETEPFVENKAAFKNEAA | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Rabbit | |
| Angiogenesis, Cancer, Cardiovascular Biology, Endosome Markers, Endothelial Cell Markers, Immunology, Inflammation, Metastasis | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE-1XLKD1 | |
| LYVE1 | |
| IgG | |
| Affinity Purified | |
| 35 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title