Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYZL6 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094491 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094491 20 μL
NB094490 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094491 Supplier Novus Biologicals Supplier No. NBP30483120UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LYZL6 Polyclonal specifically detects LYZL6 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen LYZL6
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias EC 3.2.1.17, LYC1lysozyme homolog, lysozyme-like 6, lysozyme-like protein 6, PRO1485, TKAL754, UNQ754
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-148 of human LYZL6 (NP_065159.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57151
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.