Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LZIC Antibody (OTI5E3), Biotin, Novus Biologicals™
SDP

Catalog No. NBP272561B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272561B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272561B Supplier Novus Biologicals Supplier No. NBP272561B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LZIC Monoclonal specifically detects LZIC in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen LZIC
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5E3
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias leucine zipper and CTNNBIP1 domain containing, Leucine zipper and CTNNBIP1 domain-containing protein, Leucine zipper and ICAT homologous domain-containing protein, leucine zipper domain and ICAT homologous domain containing, MGC15436, protein LZIC
Gene Symbols LZIC
Host Species Mouse
Immunogen Full length recombinant protein of human LZIC (NP_115744) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 84328.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.