Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
m-Cresol, 99%
Click to view available options
Quantity:
250 g
1000 g
2500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
m-Cresol is employed in the preparation of synthetic resins like phenol-formaldehyde resins, disinfectants, and photographic developers. It is used as industrial solvent for dissolving polymers such as polyaniline. It acts as a precursor to vitamin E and antiseptic amylmetacresol. It is used as insuline preservatives.
Solubility
Miscible with water, diethylether, chloroform and ethanol.
Notes
Hygroscopic. Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases, strong acids, aluminum and aluminum alloys.
m-Cresol is employed in the preparation of synthetic resins like phenol-formaldehyde resins, disinfectants, and photographic developers. It is used as industrial solvent for dissolving polymers such as polyaniline. It acts as a precursor to vitamin E and antiseptic amylmetacresol. It is used as insuline preservatives.
Solubility
Miscible with water, diethylether, chloroform and ethanol.
Notes
Hygroscopic. Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases, strong acids, aluminum and aluminum alloys.
Specifications
Specifications
| Melting Point | 11°C to 12°C |
| Boiling Point | 202°C to 204°C |
| UN Number | UN2076 |
| Beilstein | 506719 |
| Merck Index | 14,2579 |
| Solubility Information | Miscible with water,diethylether,chloroform and ethanol. |
| SMILES | CC1=CC(=CC=C1)O |
| Formula Weight | 108.14 |
| Percent Purity | 99% |
| Odor | Phenol-like |
| Show More |
Safety and Handling
| Hazard Category | H227-H301+H311-H314-H335 |
| Hazard Statement | GHS H Statement H301-H311-H314-H318-H227 Toxic if swallowed. Toxic in contact with skin. Causes severe skin burns and eye damage. Causes serious eye damage. Combustible liquid. |
| Precautionary Statement | P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CRESOLS, LIQUID |
| EINECSNumber | 203-577-9 |
| RTECSNumber | GO6125000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction