Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

M-CSF Antibody (007), Alexa Fluor™ 750
SDP

Catalog No. NB078033 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB078033 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB078033 Supplier Novus Biologicals Supplier No. NBP290794AF750

Rabbit Monoclonal Antibody

M-CSF Monoclonal specifically detects M-CSF in Mouse samples. It is validated for ELISA.

Specifications

Antigen M-CSF
Applications ELISA
Classification Monoclonal
Clone 7
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias colony stimulating factor 1 (macrophage), CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, M-CSF, MCSFlanimostim, MGC31930
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse M-CSF (P07141-1; Met1-Glu262).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1435
Test Specificity No cross-reactivity in ELISA with: Human CSF1 Mouse Flt3-ligand
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.