Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

M-CSF Antibody (OTI8E1), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N272601A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272601A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272601A647 Supplier Novus Biologicals Supplier No. NBP272601AF647

Mouse Monoclonal Antibody

M-CSF Monoclonal specifically detects M-CSF in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen M-CSF
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI8E1
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias colony stimulating factor 1 (macrophage), CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, M-CSF, MCSFlanimostim, MGC31930
Gene Symbols CSF1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 223-496 of human M-CSF (NP_000748) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1435.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.