Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

M-CSFR/CD115 Antibody (220), Alexa Fluor™ 488
SDP

Catalog No. NB072836 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB072836 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB072836 Supplier Novus Biologicals Supplier No. NBP290384AF488

Rabbit Monoclonal Antibody

M-CSFR/CD115 Monoclonal specifically detects M-CSFR/CD115 in Mouse samples. It is validated for ELISA.

Specifications

Antigen M-CSFR/CD115
Applications ELISA
Classification Monoclonal
Clone 220
Conjugate Alexa Fluor 488
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, Proto-oncogene c-Fms
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse M-CSFR/CD115 (NP_001032948.2; Met 1-Ser 511).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Myeloid derived Suppressor Cell, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 1436
Test Specificity No cross-reactivity in ELISA with: Human CSF1R
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.