Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

M-CSFR/CD115 Antibody (460615), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB371450 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371450 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371450 Supplier Novus Biologicals Supplier No. FAB3818Y
Add to Cart
Add to Cart

Rat Monoclonal Antibody

M-CSFR/CD115 Monoclonal specifically detects M-CSFR/CD115 in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen M-CSFR/CD115
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 460615
Conjugate DyLight 680
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, Proto-oncogene c-Fms
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse M-CSF R/CD115, Ala20-Ser511, Accession # P09581
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Myeloid derived Suppressor Cell, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 1436
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.