Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

M-Ras/R-Ras3 Antibody (18E11), Novus Biologicals™
SDP

Catalog No. NB298718 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298718 50 μL
NB298719 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298718 Supplier Novus Biologicals Supplier No. NBP32612950UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

M-Ras/R-Ras3 Monoclonal antibody specifically detects M-Ras/R-Ras3 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen M-Ras/R-Ras3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 18E11
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias FLJ42964, muscle and microspikes RAS, muscle RAS oncogene homolog, ras-related protein M-Ras, Ras-related protein R-Ras3, R-RAS3, RRAS3M-RAs
Host Species Rabbit
Immunogen A synthesized peptide derived from Human M-Ras/R-Ras3 [UniProt O14807]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 22808
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.