Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Xylylenediamine, 99%

Catalog No. p-3758986
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle

CAS: 1477-55-0 | C8H12N2 | 136.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1477-55-0
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.2
MDL Number MFCD00008119
InChI Key FDLQZKYLHJJBHD-UHFFFAOYSA-N
Synonym m-xylylenediamine, 1,3-benzenedimethanamine, 1,3-bis aminomethyl benzene, 1,3-xylylenediamine, 1,3-phenylenedimethanamine, m-xylylendiamin, 1,3-xylenediamine, mxda, m-phenylenebis methylamine, methylamine, m-phenylenebis
PubChem CID 15133
IUPAC Name [3-(aminomethyl)phenyl]methanamine
SMILES C1=CC(=CC(=C1)CN)CN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.0300g/mL
Boiling Point 265°C (745.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5700 to 1.572
Linear Formula C6H4(CH2NH2)2
Quantity 5 mL
Beilstein 13, IV, 293
Specific Gravity 1.03
Formula Weight 136.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material m-Xylylenediamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.<br
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 216-032-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.