Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

M13 bacteriophage G3P Rabbit anti-Virus, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB301763 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301763 100 μL
NB187978 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB301763 Supplier Novus Biologicals Supplier No. NBP314651100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

M13 bacteriophage G3P Polyclonal antibody specifically detects M13 bacteriophage G3P in Virus samples. It is validated for Western Blot, ELISA

Specifications

Antigen M13 bacteriophage G3P
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:5000-1:10000
Formulation 0.2um filtered solution in PBS without Preservative
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified M13 Bacteriophage coat protein G3P (P69168, Ala19-Ala130)
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.