Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
M13 bacteriophage G3P Rabbit anti-Virus, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$160.20 - $464.00
Specifications
Antigen | M13 bacteriophage G3P |
---|---|
Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
M13 bacteriophage G3P Polyclonal antibody specifically detects M13 bacteriophage G3P in Virus samples. It is validated for Western Blot, ELISASpecifications
M13 bacteriophage G3P | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Virus | |
IgG | |
Antigen and protein A Affinity-purified |
Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 | |
Polyclonal | |
Purified | |
RUO | |
Produced in rabbits immunized with purified M13 Bacteriophage coat protein G3P (P69168, Ala19-Ala130) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title