Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ M13 Sequencing Forward Primer, HPLC

Catalog No. a15637
Encompass_Preferred
Click to view available options
Quantity:
50 nmol

M13 Sequencing Forward Primer, HPLC

Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Delivery Type Single Tube, Dried-down
For Use With (Equipment) Mass Spectrometry
Description M13 (-21) Forward Sequencing Primer, 50 nmole, HPLC Purification, 900pmole Minimal Spec Yield, 5' TGTAAAACGACGGCCAGT Sequence, Delivered in a Single tube, Dried-down, Used for BigDye™ Cycle Sequencing Primers
Product Type M13 (-21) Forward Sequencing Primer
Protein Degradation Sequence 5' TGTAAAACGACGGCCAGT
Primer M13 (-21) Forward Sequencing
Quantity 50 nmol
Grade HPLC
For Use With (Application) BigDye™ Cycle Sequencing Primers
Form Solid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.