Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ M1AP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566244
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA566244 100 μL
1 options

Catalog No. PIPA566244

Supplier: Invitrogen™ PA566244

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSWADICTNLC EALQNFFSLA CSLMGPSRMS LFSLYMVQDQ HECILPFVQV KGNFARLQTC ISELRMLQRE GCFRSQGASL RLAVEDGLQQ FKQYSR Highest antigen sequence identity to the following orthologs - mouse 81%, rat 80%.

May be involved in gametogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen M1AP
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene M1AP
Gene Accession No. Q8TC57
Gene Alias C2orf65; D6Mm5e; M1AP; Meiosis 1 arrest protein; meiosis 1 arresting protein; meiosis 1 associated protein; meiosis 1-arresting protein; Meiosis 1-associated protein; RGD1563842; Spata37; spermatogenesis associated 37; spermatogenesis-associated protein 37
Gene Symbols M1AP
Host Species Rabbit
Immunogen Recombinant Human M1AP. Recombinant protein control fragment (Product #RP-106925).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130951
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.