Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ M8 B hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Description
M8 B hydrochloride is a potent and selective TRPM8 channel blocker. Blocks cold- and icilin -induced TRPM8 activation in vitro (IC50 values are 7.8 and 26.9 nM respectively). Exhibits no effect at other TRP channels (IC50 > 20 μM). Reduces body temperature in vivo.
Specifications
Specifications
Quantity | 10 mg |
Product Type | M8 B hydrochloride |
Purity | >99% |
Molecular Weight (g/mol) | 432.96 |
Inhibitors | M8 B hydrochloride |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction