Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAATS1 Antibody (OTI6G1), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB272309A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272309A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272309A48 Supplier Novus Biologicals Supplier No. NBP272309AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAATS1 Monoclonal specifically detects MAATS1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen MAATS1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6G1
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AAT1, AAT-1, AAT1alpha, AAT1-alpha, AAT1DKFZp781A2221, AMY-1-associating protein expressed in testis 1, C3orf15, chromosome 3 open reading frame 15, MYCBP/AMY-1-associated, testis expressed 1, MYCBP-binding protein, SPATA26
Gene Symbols MAATS1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-313 of human C3orf15 (NP_203528) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 89876.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.