Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAB21L3 Antibody (OTI1F6), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP272563C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272563C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272563C Supplier Novus Biologicals Supplier No. NBP272563C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAB21L3 Monoclonal specifically detects MAB21L3 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen MAB21L3
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI1F6
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C1orf161, FLJ38716, mab-21-like 3 (C. elegans), protein mab-21-like 3, RP5-1185H19.1
Gene Symbols MAB21L3
Host Species Mouse
Immunogen Full length recombinant protein of human MAB21L3 (NP_689580) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 126868.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.