Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAbPac™ HIC-10 HPLC Column
GREENER_CHOICE

Catalog No. p-6183639
Change view
Click to view available options
Length (Metric):
10 mm
100 mm
250 mm
Diameter (Metric):
4.6 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
03-898-140 100 mm 4.6 mm
03-898-141 250 mm 4.6 mm
03-898-142 10 mm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-898-140 Supplier Thermo Scientific™ Supplier No. 088480
Add to Cart
Add to Cart

Separate monoclonal antibodies (mAbs) and related biologics with high resolution, excellent biocompatibility, and selectivity using the unique chemistry of this hydrophobic interaction column.

Designed for separation of monoclonal antibodies (mAbs) and related biologics by hydrophobic interaction, the Thermo Scientific™ MAbPac™ HIC-10 LC column is a high resolution silica column. The MAbPac HIC-10 column's unique chemistry provides high resolution, excellent biocompatibility, and selectivity that is different from other HIC columns on the market.

Unique Chemistry Designed for Optimal Selectivity of mAbs

The MAbPac HIC-10 column is based on high-purity, spherical, wide-pore (1,000Å), 5μm silica particles functionalized with proprietary alkyl amide groups. Its advanced surface bonding technology leads to excellent chemical stability, high efficiency, and unique selectivity optimized for mAbs and different from other HIC phases.
  • Excellent biocompatibility and extremely low carryover
  • High resolution and high efficiency
  • Compatible with both organic solvent and aqueous mobile phases
  • Rugged column stability
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 4.6 mm
Length (Metric) 100 mm
Max. Pressure 6000 psi (415 bar)
Particle Size 5 μm
pH 2 to 8
Pore Size 1000 Å
Quantity Each
Temperature 60°C
Stationary Phase Silica
Product Line MAbPac
Packing Material Spherical, Fully Porous, Ultrapure Silica
Column Type Hydrophobic Interaction
Phase Hydrophobic Interaction
Technique Bio-LC
Green Features FSC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.