Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAbPac™ SCX-10 RS Monoclonal Antibody Analysis Column, 5μm
GREENER_CHOICE

Catalog No. 03898147
Change view
Click to view available options
Length (Metric):
50 mm
150 mm
250 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
03-898-147 150 mm
03-898-146 50 mm
03-898-148 250 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-898-147 Supplier Thermo Scientific™ Supplier No. 088242
Add to Cart
Add to Cart

For high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants, use Thermo Scientific™ MAbPac™ SCX-10 RS Monoclonal Antibody Analysis Column, 5μm.

For high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants, use Thermo Scientific™ MAbPac™ SCX-10 RS Monoclonal Antibody Analysis Columns, 5μm. The unique nonporous pellicular resin provides the resolving power to separate mAb variants that differ by as little as one charged residue. A hydrophilic layer surrounds the polymeric beads and eliminates hydrophobic interactions with the resin, resulting in very efficient peaks. A proprietary grafted cation-exchange surface provides pH selectivity control for high-resolution separations.

Superior performance for mAb variant analysis

  • Exceptionally high resolution for mAb variants
  • Ideal for characterization and quality control assessment of mAbs
  • Well-suited for stability studies, mAb therapeutic method development, and high-throughput screening
  • Unmatched column-to-column and lot-to-lot reproducibility eliminates column variability as a concern in stability and QA/QC testing
  • Meets the regulatory requirements for biopharmaceutical characterization
  • Available in 3, 5, and 10μm particle sizes in standard and UHPLC formats. The smaller-particle-size columns provide the same resolution as the 10μm columns, but with significantly faster analysis time.
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 2.1 mm
Flow Rate 0.46 mL/min
Length (Metric) 150 mm
Max. Pressure 7000 psi (480 bar)
Particle Size 5 μm
pH 2 to 12
Pore Size Nonporous
Quantity Each
Temperature Ambient to 60°C
Stationary Phase Sulfonate
Product Line MAbPac
Packing Material Spherical, Nonporous Polymer
Column Type Ion Exchange
Phase Ion Exchange
Technique Bio-LC
Green Features FSC
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.