Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Macro H2A.2 Antibody (OTI1C2), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP27264A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27264A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27264A30 Supplier Novus Biologicals Supplier No. NBP272564AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Macro H2A.2 Monoclonal specifically detects Macro H2A.2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Macro H2A.2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1C2
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias core histone macro-H2A.2, core histone macroH2A2.2, H2A histone family, member Y2, Histone macroH2A2, macroH2A2, mH2A2
Gene Symbols MACROH2A2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 2-261 of human Macro H2A.2 (NP_061119) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 55506.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.