Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Macro H2A.2 Antibody (OTI1C2), PerCP, Novus Biologicals™
SDP

Catalog No. NB288032 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288032 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288032 Supplier Novus Biologicals Supplier No. NBP272564PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Macro H2A.2 Monoclonal antibody specifically detects Macro H2A.2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Macro H2A.2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1C2
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias core histone macro-H2A.2, core histone macroH2A2.2, H2A histone family, member Y2, Histone macroH2A2, macroH2A2, mH2A2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 2-261 of human H2AFY2 (NP_061119) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55506
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.