Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Macrophage Antibody (RM0029-11H3), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N212436PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N212436PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N212436PEC7 Supplier Novus Biologicals Supplier No. NBP212436PECY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Macrophage Monoclonal specifically detects Macrophage in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Macrophage
Applications Flow Cytometry
Classification Monoclonal
Clone RM0029-11H3
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 5.3.2.1, EC 5.3.3.12, GIFmacrophage migration inhibitory factor, GLIF, Glycosylation-inhibiting factor, L-dopachrome isomerase, L-dopachrome tautomerase, macrophage migration inhibitory factor (glycosylation-inhibiting factor), MMIF, Phenylpyruvate tautomerase
Host Species Rat
Immunogen Isolated mouse macrophages
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.