Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Macrophage Marker Antibody (5/9), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP25048200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25048200 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP25048200 Supplier Novus Biologicals Supplier No. NBP250482AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Macrophage Marker Monoclonal specifically detects Macrophage Marker in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Macrophage Marker
Applications Immunohistochemistry
Classification Monoclonal
Clone 5/9
Conjugate Alexa Fluor 700
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen A cell suspension containing osteoclasts from osteoclastomas.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.