Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Macrophage Marker Antibody (5/9), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB309344 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309344 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309344 Supplier Novus Biologicals Supplier No. NBP250482MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Macrophage Marker Monoclonal antibody specifically detects Macrophage Marker in Human samples. It is validated for Immunohistochemistry

Specifications

Antigen Macrophage Marker
Applications Immunohistochemistry
Classification Monoclonal
Clone 2024-05-09 00:00:00
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen A cell suspension containing osteoclasts from osteoclastomas.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.