Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Macrophage Marker Antibody (5/9), PerCP, Novus Biologicals™
SDP

Catalog No. NB309343 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309343 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309343 Supplier Novus Biologicals Supplier No. NBP250482PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Macrophage Marker Monoclonal antibody specifically detects Macrophage Marker in Human samples. It is validated for Immunohistochemistry

Specifications

Antigen Macrophage Marker
Applications Immunohistochemistry
Classification Monoclonal
Clone 2024-05-09 00:00:00
Conjugate PerCP
Dilution Immunohistochemistry
Formulation PBS
Host Species Mouse
Immunogen A cell suspension containing osteoclasts from osteoclastomas.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.