Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAD1L1/MAD1 Antibody (9B10), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB1205783UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1205783UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1205783UV Supplier Novus Biologicals Supplier No. NB1205783UV

Mouse Monoclonal Antibody

MAD1L1/MAD1 Monoclonal specifically detects MAD1L1/MAD1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen MAD1L1/MAD1
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 9B10
Conjugate DyLight 350
Dilution Immunocytochemistry/Immunofluorescence
Gene Alias hMAD1, MAD1 mitotic arrest deficient-like 1 (yeast), MAD1-like protein 1, MAD1yeast, homolog-like 1, Mitotic checkpoint MAD1 protein homolog, Tax-binding protein 181, tumor protein p53 inducible protein 9, TXBP181MAD1 (mitotic arrest deficient, yeast, homolog)-like 1
Gene Symbols MAD1L1
Host Species Mouse
Immunogen Recombinant full length human MAD1 protein.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, DNA Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8379
Test Specificity MAD1 (9B10)
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.