Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAD1L1/MAD1 Rabbit anti-Human, Mouse, Clone: 7O2C2, Novus Biologicals™
SDP

Catalog No. NB168198 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168198 20 μg
NB168197 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB168198 Supplier Novus Biologicals Supplier No. NBP31655320UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MAD1L1/MAD1 Monoclonal antibody specifically detects MAD1L1/MAD1 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen MAD1L1/MAD1
Applications Western Blot
Classification Monoclonal
Clone 7O2C2
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias hMAD1, MAD1 mitotic arrest deficient-like 1 (yeast), MAD1-like protein 1, MAD1yeast, homolog-like 1, Mitotic checkpoint MAD1 protein homolog, Tax-binding protein 181, tumor protein p53 inducible protein 9, TXBP181MAD1 (mitotic arrest deficient, yeast, homolog)-like 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50-150 of human MAD1L1/MAD1 (Q9Y6D9).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, DNA Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8379
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.