Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAD2L1 Antibody (OTI4D2), HRP, Novus Biologicals™
SDP

Catalog No. NBP272565H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272565H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272565H Supplier Novus Biologicals Supplier No. NBP272565H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAD2L1 Monoclonal specifically detects MAD2L1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen MAD2L1
Applications Western Blot
Classification Monoclonal
Clone OTI4D2
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias HSMAD2, MAD2 (mitotic arrest deficient, yeast, homolog)-like 1, MAD2 mitotic arrest deficient-like 1 (yeast), MAD2-like protein 1, MAD2mitotic arrest deficient, yeast, homolog-like 1, Mitotic arrest deficient 2-like protein 1, mitotic spindle assembly checkpoint protein MAD2A
Gene Symbols MAD2L1
Host Species Mouse
Immunogen Full length recombinant protein of human MAD2L1 (NP_002349) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4085.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.