Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAD2L1-binding protein Antibody (OTI3C11), PE, Novus Biologicals™
SDP

Catalog No. NB288025 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288025 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288025 Supplier Novus Biologicals Supplier No. NBP272566PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAD2L1-binding protein Monoclonal antibody specifically detects MAD2L1-binding protein in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen MAD2L1-binding protein
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3C11
Conjugate PE
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias Caught by MAD2 protein, CMT2MGC11282, dJ261G23.1, KIAA0110RP1-261G23.6, MAD2L1 binding protein, MAD2L1-binding protein
Host Species Mouse
Immunogen Full length human recombinant protein of human MAD2L1BP (NP_055443) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9587
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.