Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAdCAM-1 Antibody (683715), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB347117 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347117 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347117 Supplier Novus Biologicals Supplier No. FAB6056MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAdCAM-1 Monoclonal antibody specifically detects MAdCAM-1 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen MAdCAM-1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 683715
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias hMAdCAM-1, MACAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human MAdCAM-1, Leu21-Gln333, Accession # AAY82472
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8174
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.