Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAdCAM-1 Antibody (CA 102.2C1) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB10063277 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10063277 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10063277 Supplier Novus Biologicals Supplier No. NB10063277
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAdCAM-1 Monoclonal specifically detects MAdCAM-1 in Human samples. It is validated for ELISA, Functional, Western Blot.

Specifications

Antigen MAdCAM-1
Applications ELISA, Functional Assay, Western Blot
Classification Monoclonal
Clone CA 102.2C1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Functional
Gene Accession No. Q13477
Gene Alias hMAdCAM-1, MACAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1
Gene Symbols MADCAM1
Host Species Mouse
Immunogen Bacterially expressed human MAdCAM-1
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8174
Test Specificity NB100-63277 recognizes human MAdCAM-1 (mucosal addressin cell adhesion molecule -1), a 40kD protein expressed by vascular endothelium. Clone CA102.2C1 has been reported to block the binding of human cells expressing alpha4 Beta7 to immobilized MAdCAM-1 in vitro .
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.