Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MADD Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB134310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB134310 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB134310 Supplier Novus Biologicals Supplier No. NBP306250JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MADD Polyclonal specifically detects MADD in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen MADD
Applications Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DENNFLJ36300, Differentially expressed in normal and neoplastic cells, FLJ35600, IG20, Insulinoma glucagonoma clone 20, insulinoma-glucagonoma protein 20, KIAA0358RAB3GEP, MAP kinase-activating death domain protein, MAP-kinase activating death domain, Rab3 GDP/GTP exchange factor
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MADD fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8567
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.