Learn More
Description
Specifications
Specifications
| Antigen | MADD |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-2mg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 ug/ml |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | AAD12154 |
| Gene Alias | DENNFLJ36300, Differentially expressed in normal and neoplastic cells, FLJ35600, IG20, Insulinoma glucagonoma clone 20, insulinoma-glucagonoma protein 20, KIAA0358RAB3GEP, MAP kinase-activating death domain protein, MAP-kinase activating death domain, Rab3 GDP/GTP exchange factor |
| Gene Symbols | MADD |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
