Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MafA Antibody (BLR067G), Novus Biologicals™
SDP

Catalog No. NB000888 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB000888 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB000888 Supplier Novus Biologicals Supplier No. NBP278707
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MafA Monoclonal specifically detects MafA in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen MafA
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BLR067G
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunohistochemistry-Paraffin
Formulation Borate Buffered Saline (BBS) pH 8.2. with 0.09% Sodium Azide
Gene Alias hMafA, Pancreatic beta-cell-specific transcriptional activator, RIPE3B1, transcription factor MafA, Transcription factor RIPE3b1, V-maf musculoaponeurotic fibrosarcoma oncogene homolog A, v-maf musculoaponeurotic fibrosarcoma oncogene homolog A (avian)
Gene Symbols MAFA
Host Species Rabbit
Immunogen Immunogen was a peptide representing a region between residue 125 and 175 of human MAF bZIP transcription factor A using the numbering given in entry NP_963883.2 (Gene ID 389692).
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μL
Research Discipline Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 389692
Target Species Human, Mouse
Content And Storage Store at 2-8°C/ 1 year from date of receipt
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.