Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MafA Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309249100UL
Description
MafA Polyclonal specifically detects MafA in Human samples. It is validated for Western Blot.Specifications
MafA | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
hMafA, Pancreatic beta-cell-specific transcriptional activator, RIPE3B1, transcription factor MafA, Transcription factor RIPE3b1, V-maf musculoaponeurotic fibrosarcoma oncogene homolog A, v-maf musculoaponeurotic fibrosarcoma oncogene homolog A (avian) | |
The immunogen is a synthetic peptide directed towards the N terminal region of human MafA (NP_963883). Peptide sequence PSPGTGGGGGAGGGGGSSQAGGAPGPPSGGPGAVGGTSGKPALEDLYWMS | |
100 μg | |
Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators | |
389692 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction